Tube racks, universal
Pink, 32/32/12/4, 0,5/1,5-2,0/15/50 ml, PP, 172×50×90 mm
Catalog # DELTM-522
Supplier: DELTALAB
Jump to:
Product Details & Documents
PP, linkable to other racks.
- Versatile rack with different capacities on each of the four sides
- Alpha-numeric matrix on the four sides for better sample location
- Suitable for freezers and autoclaves
Documents
Certificates
Search by Lot NumberSpecifications
- For tubes:0,5/1,5-2,0/15/50 ml
- No. of Holes:32/32/12/4
- Colour:Pink
- Material:PP
- Seq:02
- W×D×H:172×50×90 mm