Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
sieve ring for impact blade, 0.5mm trapez. perforation 1 * 1 items
Catalog # FRIT44.1351.10
Supplier:
FRITSCH
Expand image
sieve ring for impact blade, 0.5mm trapez. perforation 1 * 1 items
Catalog # FRIT44.1351.10
Supplier:
FRITSCH