Orders LinkContactUsLinkComponent
Standard ground joints
Standard ground joints
12/21 NS, - NS, 11 mm
Catalog # QUTW1043
Standard ground joints
Catalog # QUTW1043
Standard ground joints
Standard ground joints
12/21 NS, - NS, 11 mm
Price
80,80 €
Log in for availability
1 * 1 items
Product Details & Documents
Connections, quartz glass, with standard ground joint according to DIN 12 249.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving high resistance to temperature changes.
Specifications
  • Cone size:
    12/21 NS
  • Socket size:
    - NS
  • Tube ext.Ø:
    11 mm
  • Seq:
    05