Orders LinkContactUsLinkComponent
Clamp gaskets, HelixMark®
Clamp gaskets, HelixMark®
19,05 mm
Catalog # 113-0317
Clamp gaskets, HelixMark®
Catalog # 113-0317
Clamp gaskets, HelixMark®
Clamp gaskets, HelixMark®
19,05 mm
Price
556,00 €
Log in for availability
1 * 25 items
Product Details & Documents
Silicone, platinum-cured.
  • Manufactured according to GMP in a production facility registered by the FDA for the manufacture of medical products
  • Can be sterilised by autoclaving, by ethylene oxide and via gamma radiation within the user's validation cycles and sterilisation levels
  • Meet USP Class VI requirements. Material certification and lot traceability are included in each box
These biomedical/pharmaceutical grade clamp gaskets promote crevice-free and protrusion-free processing. Contain no contaminants or leachables.
ISO 13485
Certifications: ISO 13485
Specifications
  • Ø int.:
    19,05 mm
  • Seq:
    02