Order Entry
Export
Orders LinkContactUsLinkComponent
 

Ferrules for single ferrule column connectors

Ferrule, 0,25 mm to 0,25 mm ID, 15% graphite/85% Vespel®
Catalog # VARICP4783
Supplier:   VARIAN
Ferrules for single ferrule column connectors
Catalog # VARICP4783
Supplier:  VARIAN
Ferrules for single ferrule column connectors
Ferrules for single ferrule column connectors

Ferrule, 0,25 mm to 0,25 mm ID, 15% graphite/85% Vespel®

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Description:
    Ferrule, 0,25 mm to 0,25 mm ID
  • Material:
    15% graphite/85% Vespel®
  • Seq:
    08