Order Entry
Export
Orders LinkContactUsLinkComponent
Ferrules for single ferrule column connectors

Ferrules for single ferrule column connectors

Ferrule, 0,32 mm to 0,32 mm ID, 15% graphite/85% Vespel®
Catalog # VARICP4781
Supplier:   VARIAN
Ferrules for single ferrule column connectors
Catalog # VARICP4781
Supplier:  VARIAN
Ferrules for single ferrule column connectors
Ferrules for single ferrule column connectors

Ferrule, 0,32 mm to 0,32 mm ID, 15% graphite/85% Vespel®

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Description:
    Ferrule, 0,32 mm to 0,32 mm ID
  • Material:
    15% graphite/85% Vespel®
  • Seq:
    06