Solid phase extraction cartridges, Bond Elut™ C18 OH
Straight barrel cartridge, C18 OH, 150 Å, 1 ml, 100 mg
Catalog # VARI12102020
Supplier: VARIAN
Jump to:
Product Details & Documents
Bond Elut™ C18 OH is a non-end capped version of the octadecyl bonded phases that enables the silanols on the silica surface to be more active. This low-load C18 has well-controlled silanol activity that permits the fractionation of metabolites and enhances retention of basic compounds compared to an endcapped C18. The pore size makes it a good choice for peptide extraction.
- Silanol activity permits metabolite fractionation
- Tight QC tolerances deliver batch-to-batch reproducibility
- 150 Å pore size expands retention window to higher molecular weight compounds
Specifications
- Pore size:150 Å
- Capacity:1 ml
- Description:Straight barrel cartridge
- Seq:03
- Bed weight:100 mg
- Phase:C18 OH
- Carbon load:14,9%
- Cat. No.:VARI12102020
- Endcapped:No
- Surface area:300 m²/g



