Solid phase extraction cartridges, Bond Elut™ C18 EWP
Straight barrel cartridge, C18 EWP, 500 Å, 40 µm, 1 ml, 50 mg
Catalog # VARI12102136
Supplier: VARIAN
Jump to:
Product Details & Documents
Bond Elut™ EWP is based upon standard particle size silica but with 500 Å pores to allow more efficient extraction of large molecules (>15000 MW), which are typically excluded from standard porosity silica phases.
- No exclusion of large molecules
- Good for desalting proteins
- Successful separation of proteins, peptides or nucleotides
Specifications
- Particle Size:40 µm
- Pore size:500 Å
- Capacity:1 ml
- Description:Straight barrel cartridge
- Bed weight:50 mg
- Phase:C18 EWP
- Carbon load:6,0%
- Cat. No.:VARI12102136
- Endcapped:Yes
- Surface area:80 m²/g



