Order Entry
Export
Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Bond Elut™ C18 OH

Solid phase extraction cartridges, Bond Elut™ C18 OH

Straight barrel cartridge, C18 OH, 150 Å, 40 and 120, 6 ml, 1 g
Catalog # VARI12256040
Supplier:   VARIAN
Solid phase extraction cartridges, Bond Elut™ C18 OH
Catalog # VARI12256040
Supplier:  VARIAN
Solid phase extraction cartridges, Bond Elut™ C18 OH

Straight barrel cartridge, C18 OH, 150 Å, 40 and 120, 6 ml, 1 g

Price
210,74 €
Log in for availability
1 * 30 items
Product Details & Documents

Bond Elut™ C18 OH is a non-end capped version of the octadecyl bonded phases that enables the silanols on the silica surface to be more active. This low-load C18 has well-controlled silanol activity that permits the fractionation of metabolites and enhances retention of basic compounds compared to an endcapped C18. The pore size makes it a good choice for peptide extraction.

  • Silanol activity permits metabolite fractionation
  • Tight QC tolerances deliver batch-to-batch reproducibility
  • 150 Å pore size expands retention window to higher molecular weight compounds
Specifications
  • Particle Size:
    40 and 120
  • Pore size:
    150 Å
  • Capacity:
    6 ml
  • Description:
    Straight barrel cartridge
  • Bed weight:
    1 g
  • Phase:
    C18 OH
  • Carbon load:
    14,9%
  • Cat. No.:
    VARI12256040
  • Endcapped:
    No
  • Surface area:
    300 m²/g