Order Entry
Export
Orders LinkContactUsLinkComponent
 

Solid phase extraction cartridges, Bond Elut™ C18 EWP

Straight barrel cartridge, C18 EWP, 500 Å, 40 µm, 6 ml, 1 g
Catalog # VARI12256130
Supplier:   VARIAN
Solid phase extraction cartridges, Bond Elut™ C18 EWP
Catalog # VARI12256130
Supplier:  VARIAN
Solid phase extraction cartridges, Bond Elut™ C18 EWP

Straight barrel cartridge, C18 EWP, 500 Å, 40 µm, 6 ml, 1 g

Price
230,17 €
Log in for availability
1 * 30 items
Product Details & Documents

Bond Elut™ EWP is based upon standard particle size silica but with 500 Å pores to allow more efficient extraction of large molecules (>15000 MW), which are typically excluded from standard porosity silica phases.

  • No exclusion of large molecules
  • Good for desalting proteins
  • Successful separation of proteins, peptides or nucleotides
Specifications
  • Particle Size:
    40 µm
  • Pore size:
    500 Å
  • Capacity:
    6 ml
  • Description:
    Straight barrel cartridge
  • Bed weight:
    1 g
  • Phase:
    C18 EWP
  • Carbon load:
    6,0%
  • Cat. No.:
    VARI12256130
  • Endcapped:
    Yes
  • Surface area:
    80 m²/g