Orders LinkContactUsLinkComponent
Triphenylsilane ≥99%
Glass bottle for solids, 50 g
  A11605.18
Triphenylsilane ≥99%
  A11605.18
 :  789-25-3
Triphenylsilane ≥99%
Triphenylsilane ≥99%
Glass bottle for solids, 50 g
 :  789-25-3
171,00 €
1 * 50 g
Purity: 99% CAS No.: 789-25-3 Molecular Formula: (C6H5)3SiH Formula Weight: 260.41 Flash Point: 76°(169°F) Melting Point: 42-44° Boiling Point: 159-163°/3mm Beilstein Reference: 978182 MDL No.: MFCD00003003
  •  :789-25-3
  •  :C₁₈H₁₆Si
  •  :MFCD00003003
  •  :260.41 g/mol
  •  :159…163 °C (3 mmHg)
  •  :42…44 °C
  •  :1.152 g/cm³ (20 °C)
  •  :76 °C (169 °F)
  •  :Ambient
  •  :212-333-0
  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g