Order Entry
Export
Orders LinkContactUsLinkComponent
 

Cyclohexane, Sigma-Aldrich®

Glass bottle, 1 L
Catalog # SIAL227048-1L
Flammable
Health
Irritant
Environment
Cyclohexane, Sigma-Aldrich®
Catalog # SIAL227048-1L
Supplier:  
CAS Number:  110-82-7
Cyclohexane, Sigma-Aldrich®
Cyclohexane, Sigma-Aldrich®

Glass bottle, 1 L

CAS Number:  110-82-7
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
See more alternatives
Product Details & Documents
Synonyms:  Hexahydrobenzene, Hexamethylene
  • CAS Number:110-82-7
  • Formula:C₆H₁₂
  • MDL Number:MFCD00003814
  • Molecular Weight:84.16 g/mol
  • Merck Index:13,02752
  • Boiling Pt:81 °C (1013 hPa)
  • Melting Pt:7 °C
  • Density:0.7785 g/cm³ (20 °C)
  • Flash Pt:–18 °C
  • Storage Temperature:Ambient
  • UN:1145
  • ADR:3,II
  •  :M-Clarity™ quality level = MQ200
  • EINECS:203-806-2
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications