Orders LinkContactUsLinkComponent
Push-in caps for tubes
Push-in caps for tubes
LDPE, Caps, Ø 3 mm, Natural
Catalog # SEMA1435
Supplier:   SEMADENI
Push-in caps for tubes
Catalog # SEMA1435
Supplier:  SEMADENI
Push-in caps for tubes
Push-in caps for tubes
LDPE, Caps, Ø 3 mm, Natural
Price
5,60 €
Log in for availability
1 * 100 items
Product Details & Documents
LDPE, translucent.
Caps with rim for sealing test tubes, flasks and bottles.
Specifications
  • Colour:
    Natural
  • Description:
    Caps, Ø 3 mm
  • Material:
    LDPE
  • Seq:
    01