Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Media for Microbiology
Additives and Supplements for Microbiology
Blood
Print
Blood
Sheep Blood Defibrinated, 25 mL
Catalog # OXOISR0051B
Certificates
Supplier:
THERMO FISHER DIAGNOSTICS
Blood
Catalog # OXOISR0051B
Supplier:
THERMO FISHER DIAGNOSTICS
CAS Number:
Certificates
Return to Top
Blood
Sheep Blood Defibrinated, 25 mL
Specifications
Jump to:
Specifications
CAS Number:
Price
16,30 €
Log in
for availability
1 * 25 mL
-
+
Other Product Family Options Available
Return Policy
Product Details & Documents
Documents
Certificates
Search by Lot Number
Datasheet
OXOISR0048C_TDS_EN
Specifications
Specifications
Pk:
25 mL
Description:
Sheep Blood Defibrinated
Specifications