Naratriptan Related Compounds, Reference Standard
Naratriptan Hydrochloride, USP reference standard, Vial, 125 mg
Catalog # 1457469.
Supplier: USP
Jump to:
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications
Specifications
- Pack type:Vial
- Pk:125 mg
- Standards:USP reference standard
- Description:Naratriptan Hydrochloride
- Cat. No.:1457469.
- Formula:C₁₇H₂₆ClN₃O₂S
- Description:Naratriptan Hydrochloride
- CAS No.:143388-64-1



