Order Entry
Export
Orders LinkContactUsLinkComponent
Desogestrel Related Compounds, Reference Standard

Desogestrel Related Compounds, Reference Standard

Desogestrel Related Compound C, USP reference standard, Vial, 25 mg
Catalog # 1173268.
Supplier:   USP
Desogestrel Related Compounds, Reference Standard
Catalog # 1173268.
Supplier:  USP
CAS Number:  
Desogestrel Related Compounds, Reference Standard

Desogestrel Related Compound C, USP reference standard, Vial, 25 mg

CAS Number:  
Price
1 240,00 €
Log in for availability
1 * 25 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications

Specifications

  • Pack type:
    Vial
  • Pk:
    25 mg
  • Standards:
    USP reference standard
  • Description:
    Desogestrel Related Compound C
  • Cat. No.:
    1173268.
  • Formula:
    C₂₂H₂₈O₂
  • Description:
    Desogestrel Related Compound C
  • Synonym(s):
    13-Ethyl-11-methylene-18,19-dinor-17α-pregn-4-en-20-yn-17-ol-3-one
  • CAS No.:
    54048-10-1

Specifications