Peroxidase substrates, TrueBlue™
TrueBlue™ peroxidase substrate, 1×10 ml, sufficient to stain approximately 25 - 50 slides, 10 mL
Catalog # KPLI5510-0052
Supplier: SERACARE LIFE SCIENCES
Peroxidase substrates, TrueBlue™
Catalog # KPLI5510-0052
Supplier: SERACARE LIFE SCIENCES
CAS Number:
Jump to:
Product Details & Documents
KPL TrueBlue™ peroxidase substrate is a highly sensitive chromagenic substrate for visualisation of horseradish peroxidase-labeled reporter reagents.
- Provides a brilliant blue specific stain which provides excellent contrast with red counterstains
- Free of known carcinogens
- Single component, ready to use, liquid substrate
- No mixing is required
KPL TrueBlue™ peroxidase substrate is a buffered solution containing 3,3',5,5'-tetramethylbenzidine (TMB) and H2O2. It is optimised for use with cellular preparations and can be used for immunoblotting procedures, but not for microwell ELISA or other applications requiring a soluble reaction product.
Caution: Store at room temperature (22 to 28 °C). TrueBlue is stable for up to 30 months from the date of manufacture.
Specifications
Specifications
- Pk:10 mL
- Description:TrueBlue™ peroxidase substrate, 1×10 ml, sufficient to stain approximately 25 - 50 slides



