0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
BUFFER TRAY 36 (2ML VIALS) 1 * 1 items
BECL144390
:
Beckman Coulter
BUFFER TRAY 36 (2ML VIALS) 1 * 1 items
BECL144390
:
Beckman Coulter