Order Entry
Export
Orders LinkContactUsLinkComponent
Acetone ≥99.5% ACS

Acetone ≥99.5% ACS

Glass bottle, 1 L
  E646-1L
 
 
Acetone ≥99.5% ACS
  E646-1L
 :  67-64-1
Acetone ≥99.5% ACS
Acetone ≥99.5% ACS

Glass bottle, 1 L

 :  67-64-1
 
94,10 €
   
1 * 1 L
 
 :  2-Propanone, DMK, Propanone, Dimethyl ketone
  •  :67-64-1
  •  :CH₃COCH₃
  •  :MFCD00008765
  •  :58.08 g/mol
  •  :13,00067
  •  :56.2 °C (1013 hPa)
  •  :–95.4 °C
  •  :0.792 g/cm³ (20 °C)
  •  :<–20 °C (closed cup)
  •  :Ambient
  •  :200-662-2
  •  :1090
  •  :3,II
  •  :01-2119471330-49
 

 

  • Abs.@330 nm:
    <1
  • Abs.@340 nm:
    ≤0.1
  • Abs.@350 nm:
    ≤0.02
  • Abs.@400 nm:
    ≤0.01
  • Aldehyde:
    ≤0.002%
  • Color (APHA):
    ≤10
  • Isopropyl Alcohol:
    ≤0.05%
  • Methanol:
    ≤0.05%
  • Purity:
    ≥99.5%
  • Residue After Evaporation:
    ≤0.001%
  • Solubility (50%, CO₂ free Water):
    Pass
  • Substances Reducing Permanganate Test:
    Pass
  • Titrable Acid:
    ≤0.0003 meq/g
  • Titrable Base:
    ≤0.0006 meq/g

 

  • Pack type:
    Glass bottle
  • Pk:
    1 L