Poly-L-lysine hydrobromide
Product Details & Documents
Poly-L-lysine is a positively charged amino acid polymer with approximately one HBr per lysine residue. The hydrobromide allows the poly-L-lysine to be in a crystalline form soluble in water. A small amount of product may be found in the beta structure because the HBr interferes with hydrogen bonding between amino and either the carboxyl groups or N or O containing moieties.
Poly-L-lysine polymers can be used in promoting cell adhesion to solid substrates, conjugation to methotrexate for increased drug transport, microencapsulation of islets, cell microencapsulation technology, microarray glass slide coating, and chromosomal preparations. Lower molecular weight poly-L-lysine (30,000-70,000) is less viscuous in solution, but higher molecular weight versions provide more attachment sites per molecule.
This product is supplied as a 0.10% (w/v) solution in water. Coating slides with this solution should be followed with a 5 minute incubation and drying the slides at room temperature. Coated slides will be stable for one year if protected from dust.
Documents
Merck The Color of Tissue Diagnostics
Product Literature
Specifications
Specifications
- Pack type:Plastic bottle
- Pk:100 mL
- Cat. No.:P8920-100ML
- M-Clarity quality level:MQ200
- Molecular formula:(C₆H₁₃N₂OBr)n
- Protein/peptide name:Poly-L-lysine hydrobromide
- Storage conditions:Ambient
- Formulation:Solution in water
- MDL No.:MFCD00237230
- Concentration:0.1 % w/v
- Molecular weight:150000 - 300000
- CAS No.:25988-63-0



