Orders LinkContactUsLinkComponent
Nitrogen receivers (schlenk Tubes)
Nitrogen receivers (schlenk Tubes)
10 ml, - NS, 14/23 NS
Catalog # 201-1161
Supplier:   Lenz Laborglas
Nitrogen receivers (schlenk Tubes)
Catalog # 201-1161
Supplier:  Lenz Laborglas
Nitrogen receivers (schlenk Tubes)
Nitrogen receivers (schlenk Tubes)
10 ml, - NS, 14/23 NS
Price
60,00 €
Log in for availability
1 * 1 items
Product Details & Documents
DURAN®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With 14/23 standard ground sockets/cones and standard ground side stopcock with tube connector.
Documents
Specifications
  • Cone size:
    - NS
  • Socket size:
    14/23 NS
  • Capacity:
    10 ml