Cyclohexane, Sigma-Aldrich®
Glass bottle, 2 L
Catalog # SIAL227048-2L
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Cyclohexane, 99.5%, Extra Dry, AcroSeal® 1 * 2,5 L Catalog #326830025.
Product 326830025. is similar to SIAL227048-2L but manufacturer and specifications may vary.
Product Details & Documents
Synonyms: Hexahydrobenzene, Hexamethylene
- CAS Number:110-82-7
- Formula:C₆H₁₂
- MDL Number:MFCD00003814
- Molecular Weight:84.16 g/mol
- Merck Index:13,02752
- Boiling Pt:81 °C (1013 hPa)
- Melting Pt:7 °C
- Density:0.7785 g/cm³ (20 °C)
- Flash Pt:–18 °C
- Storage Temperature:Ambient
- UN:1145
- ADR:3,II
- :M-Clarity™ quality level = MQ200
- EINECS:203-806-2
Specifications
Specifications
- Pack type:Glass bottle
- Pk:2 L



