Sudan III
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 85-86-9 Molecular Formula: C22H16N4O Formula Weight: 352.40 Melting Point: ca 200° dec. Beilstein Reference: 931185 Merck Reference: 14,8884 MDL No.: MFCD00003905
Synonyms: Solvent Red 23, Sudan® III (Red), Scarlet B, Sudan Red III, Cerasin Red, Sudan Red BK, Sudan Red B, Fat Ponceau G, Sudan G, D and C Red No. 17, Oil red, Tony Red, Oil scarlet, Organol® red BS, 1-(4-(Phenylazo)phenylazo)-2-naphthol, Sudan V, 1-((4-(Phenyldiazenyl)phenyl)diazenyl)naphthalen-2-ol
- CAS Number:85-86-9
- Formula:C₂₂H₁₆N₄O
- MDL Number:MFCD00003905
- Molecular Weight:352.4 g/mol
- Merck Index:14,8884
- Melting Pt:∼ 200 °C (dec.)
- Storage Temperature:Ambient
- EINECS:201-638-4
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



