Buffer solutions as per DIN 19266-01: 2000
Buffer solution, STAPL-4/7/9, package with 10×6 glass ampoules with each 20 ml, 4,01, 6,87, 9,18, ±0,01, 1 KIT
Catalog # WTWA109020
Supplier: WTW
Jump to:
Product Details & Documents
These standard buffer solutions (work reference buffer solutions) are widely used in precision applications.
- Traceably certified to PTB/NIST and therefore safe for use
- Every bottle is labeled with their own lot number with an expiration date
: DIN 19266-01: 2000.
Specifications
Specifications
- Pk:1 KIT
- Description:Buffer solution, STAPL-4/7/9, package with 10×6 glass ampoules with each 20 ml
- pH accuracy:±0,01
- pH value:4,01, 6,87, 9,18



