17-Octadecynoic acid
Glass bottle for solids, 1 mg
Catalog # CAYM90270-1
Supplier: Cayman Chemical
Jump to:
Product Details & Documents
- CAS Number:34450-18-5
- Formula:C₁₈H₃₂O₂
- Molecular Weight:280.45 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 mg



