Erucic acid
Glass bottle for solids, 50 mg
Catalog # CAYM90175-50
Supplier: Cayman Chemical
Jump to:
Product Details & Documents
Synonyms: (Z)-Docos-13-enoic acid
- CAS Number:112-86-7
- Formula:C₂₂H₄₂O₂
- MDL Number:MFCD00063188
- Molecular Weight:338.58 g/mol
- EINECS:204-011-3
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:50 mg



