Linoleic acid
Glass bottle for solids, 100 mg
Catalog # CAYM90150-100
Supplier: Cayman Chemical
Jump to:
Product Details & Documents
Synonyms: (9Z,12Z)-Octadeca-9,12-dienoic acid, cis,cis-9,12-Octadecadienoic acid, 9Z,12Z-Octadecadienoic acid
- CAS Number:60-33-3
- Formula:C₁₈H₃₂O₂
- MDL Number:MFCD00064241
- Molecular Weight:280.45 g/mol
- Merck Index:13,05527
- Boiling Pt:360 °C (1013 hPa)
- Melting Pt:–5 °C
- Density:0.9 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-470-9
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg



