Resveratrol
Glass bottle for solids, 100 mg
Catalog # CAYM70675-100
Supplier: Cayman Chemical
Jump to:
Product Details & Documents
Synonyms: trans-3,4,5-Trihydroxystilbene, (E)-5-(4-Hydroxystyryl)benzene-1,3-diol
- CAS Number:501-36-0
- Formula:C₁₄H₁₂O₃
- MDL Number:MFCD00133799
- Molecular Weight:228.25 g/mol
- Merck Index:12,08326
- Melting Pt:260 °C
- Density:1.36 g/cm³ (20 °C)
- Storage Temperature:Freezer
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg



