Enterolactone ELISA kit
Product Details & Documents
A competitive assay for measurement of enterolactone in plasma, urine, and other biological sample matrices.
- Measure enterolactone in plasma, urine, and other sample matrices
- Assay 24 samples in triplicate or 36 samples in duplicate
- Measure enterolactone levels down to 70 pg/ml
- Colorimetric reading at 405 to 420 nm
Enterolactone is a mammalian lignan with an estrogen-like diphenolic structure. It is produced by intestinal bacteria from two plant precursors (matairesinol and secoisolariciresinol) obtained in the diet. Enterolactone and other lignans and phytoestrogens have been associated with a reduced risk of acute coronary events, hormone-dependent cancers, and possibly osteoporosis. Several studies have suggested that serum enterolactone may serve as a biomarker of a healthy, high-fiber diet. Cayman Chemical’s Enterolactone ELISA kit is a competitive assay that utilizes a standard curve ranging from 15,6 to 2000 pg/ml. The assay has a range from 15,6 to 2500 pg/ml and a sensitivity (80% B/B0) of approximately 70 pg/ml.
Delivery information: Kit contains antiserum, tracer, standard, ELISA and wash buffer concentrate; polysorbate 20, plate and cover sheet, Ellman's reagent, tracer and antiserum dye.
Documents
Specifications
Specifications
- Pk:96 Tests
- Description:Enterolactone ELISA kit, 96 strip wells
- Cat. No.:CAYM500520-96
- Developmental time:90 - 120 min
- Sensitivity:70 pg/ml
- Detection range:15,6 - 2500 pg/ml
- Incubation time:18 hr



