Order Entry
Export
Orders LinkContactUsLinkComponent
(±)-α-Lipoic acid

(±)-α-Lipoic acid

Glass bottle for solids, 5 g
Catalog # CAYM10005728-5
Supplier:   Cayman Chemical
(±)-α-Lipoic acid
Catalog # CAYM10005728-5
Supplier:  Cayman Chemical
CAS Number:  1077-28-7
(±)-α-Lipoic acid

Glass bottle for solids, 5 g

CAS Number:  1077-28-7
Price
57,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  5-(1,2-Dithiolan-3-yl)pentanoic acid, (±)​-​1,2-​Dithiolane-​3-pentanoic acid, 5-(1,2)Dithiolan-3-yl-pentanoic acid, DL-α-Lipoic acid, DL-Thioctic acid (oxidized), DL-6,8-Thioctic acid, Lip(S2), 6,8-Dithiooctanoic acid
  • CAS Number:1077-28-7
  • Formula:C₈H₁₄O₂S₂
  • MDL Number:MFCD00005474
  • Molecular Weight:206.33 g/mol
  • EINECS:214-071-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications