(±)-α-Lipoic acid
Glass bottle for solids, 10 g
CAYM10005728-10
: 5-(1,2-Dithiolan-3-yl)pentanoic acid, (±)-1,2-Dithiolane-3-pentanoic acid, 5-(1,2)Dithiolan-3-yl-pentanoic acid, DL-α-Lipoic acid, DL-Thioctic acid (oxidized), DL-6,8-Thioctic acid, Lip(S2), 6,8-Dithiooctanoic acid
- :1077-28-7
- :C₈H₁₄O₂S₂
- :MFCD00005474
- :206.33 g/mol
- :214-071-2
- Pack type:Glass bottle for solids
- Pk:10 g