Order Entry
Export
Orders LinkContactUsLinkComponent
Luteolin

Luteolin

Glass bottle for solids, 100 mg
Catalog # CAYM10004161-100
Irritant
Supplier:   Cayman Chemical
Luteolin
Catalog # CAYM10004161-100
Supplier:  Cayman Chemical
CAS Number:  491-70-3
Luteolin

Glass bottle for solids, 100 mg

CAS Number:  491-70-3
Price
145,00 €
Log in for availability
1 * 100 mg
Product Details & Documents
Synonyms:  2-(3,4-Dihydroxyphenyl)-5,7-dihydroxy-4-benzopyrone, Natural Yellow 2, 2-(3,4-Dihydroxyphenyl)-5,7-dihydroxy-4H-chromen-4-one, 3',4',5,7-Tetrahydroxyflavone
  • CAS Number:491-70-3
  • Formula:C₁₅H₁₀O₆
  • MDL Number:MFCD00017309
  • Molecular Weight:286.24 g/mol
  • EINECS:207-741-0

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg

Specifications