Orders LinkContactUsLinkComponent
Biopsy processing/embedding cassettes, Histosette ® I M499
Biopsy processing/embedding cassettes, Histosette ® I M499
Green
Catalog # 720-0944
Supplier:   Simport Scientific
Biopsy processing/embedding cassettes, Histosette ® I M499
Catalog # 720-0944
Supplier:  Simport Scientific
Biopsy processing/embedding cassettes, Histosette ® I M499
Biopsy processing/embedding cassettes, Histosette ® I M499
Green
Price
246,00 €
Log in for availability
1 * 1.500 items
Product Details & Documents
Acetal polymer. Disposable cassettes similar to M498 series but specially designed to hold biopsy specimens during the embedding process.
  • No need for biopsy pads
  • Square openings (1 mm) to maximise fluid exchange and to ensure proper drainage
  • Integral lid
  • 45° angle writing area to enable use with some automated cassette-labelling instruments
Specifications
  • Colour:
    Green