Biopsy processing/embedding cassettes, Histosette ® I M499
Pink
720-0943
Acetal polymer. Disposable cassettes similar to M498 series but specially designed to hold biopsy specimens during the embedding process.
- No need for biopsy pads
- Square openings (1 mm) to maximise fluid exchange and to ensure proper drainage
- Integral lid
- 45° angle writing area to enable use with some automated cassette-labelling instruments
- Colour:Pink