Tissue processing/embedding cassettes, Histosette® II, M492
Purple
Catalog # 720-0906
Supplier: Simport Scientific
Tissue processing/embedding cassettes, Histosette® II, M492
Catalog # 720-0906
Supplier: Simport Scientific
Jump to:
Product Details & Documents
Acetal polymer.
- Resistant to the chemical action of histological solvents
- Flow-through slots maximise fluid exchange
- Integral lid
- Anterior writing area at 45° angle to enable use with leading cassette labelling instruments
Specifications
- Colour:Purple



