Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Speedy-Diff-Färbung A 1 * 500 mL
Speedy-Diff-Färbung A 1 * 500 mL
Print
Speedy-Diff-Färbung A 1 * 500 mL
Catalog # CTEH61079/5
Supplier:
CLIN TECH
Speedy-Diff-Färbung A 1 * 500 mL
Catalog # CTEH61079/5
Supplier:
CLIN TECH
CAS Number:
Expand image
Return to Top
Speedy-Diff-Färbung A 1 * 500 mL
Add to cart
CAS Number:
Price
49,40 €
Log in
for availability
Unit of Measure
1 * 500 mL
-
+
Add to cart
Return Policy
Product Details & Documents