VWR®, Transfer Pipettes, High Performance
6,0 ml, 22, 3,5 ml, 5 mm, 230 mm, 1,5 ml, +, 10 per bag
Supplier: VWR Collection
Jump to:
Product Details & Documents
LDPE, disposable. These unbreakable 'all in one' pipettes eliminate the potential hazard of broken glass and exposure to infectious materials. The low affinity surface reduces the loss of cells and valuable proteins due to binding. They work well whenever there is a need for quick, safe transfer of fluids.
- Non toxic and inert to biological fluids and most acids
- Can be heat-sealed and frozen at very low temperatures such as in liquid nitrogen
- Available sterile or non sterile
- Sterility assurance level is SAL 10⁻⁶
: Sterile pipettes are packed in medical grade paper and non toxic film.
Specifications
- Packed:10 per bag
- Sterile:+
- Capacity:6,0 ml
- Length:230 mm
- Seq:13
- Drops per ml:22
- Graduation:1,5 ml
- Stem Ø:5 mm
- Bulb draw:3,5 ml



