Orders LinkContactUsLinkComponent
Labels for bottles
Labels for bottles
Labels for distilled water, Blue, 130×35 mm
Catalog # KART5010
Supplier:   KARTELL
Labels for bottles
Catalog # KART5010
Supplier:  KARTELL
Labels for bottles
Labels for bottles
Labels for distilled water, Blue, 130×35 mm
Price
461,00 €
Log in for availability
1 * 100 items
Product Details & Documents
Transparent, punched labels for use with wash bottles and plastic or glass reagent bottles.
  • Complete with risk assessment numbers
  • Recommended by the NFPA (National Fire Protection Association)
Specifications
  • Colour:
    Blue
  • Description:
    Labels for distilled water
  • Dimensions:
    130×35 mm