Sample cups for analysers, ratiolab®
For Hitachi H1 analysers, 3 ml
RATI5700401
: RATIOLAB
Made of clear PS.
- Good chemical resistance
- Good temperature resistance
Analysis beakers offer the highest precision for use with robotic analytical systems.
- Capacity:3 ml
- Description:For Hitachi H1 analysers
- Seq:03