Order Entry
Export
Orders LinkContactUsLinkComponent
 

Probes for 845

Probe, waterproof, TC type K, −60...+1000 °C, Class 1 (EN 60584-2), 300 mm, 1,5×300 mm
  620-1246
 :   Testo
Probes for 845
  620-1246
 :  Testo
Probes for 845
Probes for 845

Probe, waterproof, TC type K, −60...+1000 °C, Class 1 (EN 60584-2), 300 mm, 1,5×300 mm

 
112,00 €
   
1 * 1 items
 
  • Range:
    −60...+1000 °C
  • Description:
    Probe, waterproof, TC type K
  • Length:
    300 mm
  • Seq:
    06
  • Accuracy:
    Class 1 (EN 60584-2)
  • Application:
    Immersion/penetration
  • Ø×L:
    1,5×300 mm