Orders LinkContactUsLinkComponent
VWR®, Cleanroom Mops
VWR®, Cleanroom Mops
Tubular knit mop, ribbed
Catalog # 129-0149
Supplier:   VWR Collection
VWR®, Cleanroom Mops
Catalog # 129-0149
Supplier:  VWR Collection
VWR®, Cleanroom Mops
VWR®, Cleanroom Mops
Tubular knit mop, ribbed
Price
716,00 €
Log in for availability
1 * 12 items
Product Details & Documents
100% polyester string mops. Double clear poly bagged for cleanroom use. Good chemical sorbancy. Ideal for application of cleaning solutions on floors in critical environments.
  • The sterile tubular knit mop, 129-0148, has a sterility assurance level of 10⁻⁶
  • The ribbed tubular knit mop offers the best sorbancy with chemical
Suitable for use in Class 10, ISO 4, M2.5 environments or below.
Specifications
  • Description:
    Tubular knit mop, ribbed
  • Seq:
    03