VWR®, Cleanroom Mops
Tubular knit mop
129-0147
100% polyester string mops. Double clear poly bagged for cleanroom use. Good chemical sorbancy. Ideal for application of cleaning solutions on floors in critical environments.
- The sterile tubular knit mop, 129-0148, has a sterility assurance level of 10⁻⁶
- The ribbed tubular knit mop offers the best sorbancy with chemical
Suitable for use in Class 10, ISO 4, M2.5 environments or below.
- Description:Tubular knit mop
- Seq:01