Vu-tight direct injection liners
Cyclo Vu-tight direct injection liner, with wool, Siltek® deactivated
Catalog # REST20788-214.5
Supplier: Restek
Jump to:
Product Details & Documents
These Vu-tight Direct injection (DI) liners are suitable for 6,35 mm packed injection port conversion. They are available in two designs and fit 0,32 and 0,53 mm ID capillary columns (column outer diameter from 0,5 mm to 0,8 mm).
- Visually observe the press-tight connection between the column end and liner
- Slotted top prevents obstruction of carrier gas flow
- Operate in the direct injection mode
Vu-tight DI liners can easily be packed with wool for dirty samples and cyclo Vu-Tight DI liners are ideal for dirty samples and spiral bore prevents non-volatile residue from contaminating the column.
Specifications
- Description:Cyclo Vu-tight direct injection liner, with wool, Siltek® deactivated
- Seq:17