Organic reference standards, Carbazole
Carbazole, 1000 µg/ml in methylene chloride, 1 ml/ampoule, 1 mL
Catalog # REST31836
Supplier: Restek
Jump to:
Product Details & Documents
Ordering information: Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis and material safety data sheet.
Specifications
Specifications
- Concentration:1000 µg/ml in methylene chloride
- Pk:1 mL
- Description:Carbazole
- Pk Info:1 ml/ampoule
- Cat. No.:REST31836
- Formula:C₁₂H₉N
- Description:Carbazole
- Melting point:243…246 °C (lit.)
- CAS No.:86-74-8



