Orders LinkContactUsLinkComponent
Accessories for solid phase extraction cartridges, Resprep®
Accessories for solid phase extraction cartridges, Resprep®
Frits, PE, 20 µm, for 1 ml tubes (6 mm)
  REST26016
 :   Restek
Accessories for solid phase extraction cartridges, Resprep®
  REST26016
 :  Restek
Accessories for solid phase extraction cartridges, Resprep®
Accessories for solid phase extraction cartridges, Resprep®
Frits, PE, 20 µm, for 1 ml tubes (6 mm)
Inquire For Price
Log in for availability
1 * 100 items
Resprep tubes, frits, caps, and connectors for method development needs.
Specifications
  • Description:
    Frits, PE, 20 µm, for 1 ml tubes (6 mm)
  • Seq:
    07