Order Entry
Export
Orders LinkContactUsLinkComponent
 

Packing material for columns, Res-Sil

Packing material, 2% Carbowax 1540 on Res-Sil C, mesh size 80/100, temperature limit 150 °C, 1 g
Catalog # REST25044
Supplier:   Restek
Packing material for columns, Res-Sil
Catalog # REST25044
Supplier:  Restek
CAS Number:  
Packing material for columns, Res-Sil
Packing material for columns, Res-Sil

Packing material, 2% Carbowax 1540 on Res-Sil C, mesh size 80/100, temperature limit 150 °C, 1 g

CAS Number:  
Price
88,15 €
Log in for availability
1 * 1 g
Product Details & Documents

These bonded silica packings with n-octane or cyanopropyl (OPN) functional groups yield faster separations of C1 to C4 hydrocarbons, higher thermal stability, shorter conditioning times, and longer lifetimes.

  • Unique separation of saturated and unsaturated hydrocarbons
  • Innovative bonding chemistry for batch-to-batch reproducibility

Each production batch of Res-Sil C packing is tested with a complex hydrocarbon mixture to meet demanding retention time and retention index specifications.

Specifications

Specifications

  • Pk:
    1 g
  • Description:
    Packing material, 2% Carbowax 1540 on Res-Sil C, mesh size 80/100, temperature limit 150 °C

Specifications