Precision sampling valves for vials, Mininert®
Mininert® precision sampling valves, for screw-thread vials, 20 mm/400 thread
Catalog # REST24903
Supplier: Restek
Jump to:
Product Details & Documents
Mininert® valves are very convenient for repetitive sampling and limit content exposure to the silicon septum. The crimp-top valve for 20 mm ID glassware slides into the neck of the vial.
- Available for screw cap and crimp top vials
- Threaded flange used to secure tight fit
Specifications
- Description:Mininert® precision sampling valves, for screw-thread vials, 20 mm/400 thread
- Seq:04