Order Entry
Export
Orders LinkContactUsLinkComponent
Screw thread step vials, 13/425, WISP 48

Screw thread step vials, 13/425, WISP 48

Step vial with white graduated marking spot, 4,00 ml, 15×45 mm, Glass, Amber, No
Catalog # REST24656
Supplier:   Restek
Screw thread step vials, 13/425, WISP 48
Catalog # REST24656
Supplier:  Restek
Screw thread step vials, 13/425, WISP 48
Screw thread step vials, 13/425, WISP 48

Step vial with white graduated marking spot, 4,00 ml, 15×45 mm, Glass, Amber, No

Price
73,00 €
Log in for availability
1 * 100 items
Product Details & Documents

Screw-thread step vials.

Specifications
  • Cap included:
    No
  • Ø×H:
    15×45 mm
  • Capacity:
    4,00 ml
  • Colour:
    Amber
  • Description:
    Step vial with white graduated marking spot
  • Material:
    Glass
  • Seq:
    03