Orders LinkContactUsLinkComponent
Crimp top vials with inserts
Vials with 100 µl glass insert, 2,00 ml, Plastic, Clear, No
  REST24653
 :   Restek
Crimp top vials with inserts
  REST24653
 :  Restek
Crimp top vials with inserts
Crimp top vials with inserts
Vials with 100 µl glass insert, 2,00 ml, Plastic, Clear, No
295,00 €
1 * 100 items
Crimp-top plastic vial with 100 µl glass or PP fused insert.
  • Cap included:
    No
  • Capacity:
    2,00 ml
  • Colour:
    Clear
  • Description:
    Vials with 100 µl glass insert
  • Material:
    Plastic