Order Entry
Export
Orders LinkContactUsLinkComponent
Vacuum gauges

Vacuum gauges

Vacuum gauge, −30 to 0 Hg, Ø 51 mm, 3,175 mm NTP fitting
Catalog # REST24269
Supplier:   Restek
Vacuum gauges
Catalog # REST24269
Supplier:  Restek
Vacuum gauges
Vacuum gauges

Vacuum gauge, −30 to 0 Hg, Ø 51 mm, 3,175 mm NTP fitting

Price
199,38 €
Log in for availability
1 * 1 items
Product Details & Documents

These vacuum gauges are made of stainless steel wetted surfaces with rear mount.

Suitable for passive air sampling kits.

Specifications
  • Description:
    Vacuum gauge, −30 to 0 Hg, Ø 51 mm, 3,175 mm NTP fitting
  • Seq:
    01